Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Simplify sampling with easy access
Brand:  Thermo Scientific™ 840-278100
 View more versions of this product
Product Code. 15570592
  • 365.00 € / Each
Explore more special offers
Description
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.
Get the most from your measurments with accessories for the GENESYS Vis/UV-Vis Spectrophotometer family. Simplify sampling with easy access, flexibility and choices to fit your varied testing needs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sample holder for film/filter accommodates filters/lenses up to 50 mm long, 80 mm tall and 6 mm thick. Comes assembled in a removable sample compartment liner.
Measuring transmitting solid samples such as glass or plastic panels, optical filters and specialty films on glass or plastic substrates.
Filter/lenses up to 50 mm long x 80 mm tall x 6 mm thick
Sample Holder for Film/Filter
GENESYS 30/40/50 Spectrophotometers
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers > Sample holder for film/filter