missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Single-Use Flow Tube Sensors

Build a complete, single-use flow sensor system that can be customized to any desired application

Brand:  Masterflex™ 24507-30

 View more versions of this product

Product Code. 16669872

  • 281.00 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Modular interchangeable flow sensor systems with single-use design
  • Accurate measurement readings at low flow: ±1%
  • Low operating cost, disposable flow tubes for optimal contamination-free production
  • Ideal for single-use applications, but also suitable for long-term measurement
  • Flow tubes and turbine sensors are produced and tested under cleanroom conditions
Specifications

Specifications

PFA/Clip Mount
80°C
None
Flow Sensor
176°F
0.06 to 2 LPM
5-24 VDC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers