Learn More
Masterflex™ Single-Use Flow Tube Sensors
Build a complete, single-use flow sensor system that can be customized to any desired application
274.00 € - 890.00 €
Specifications
Max. Operating Temperature (English) | 176°F |
---|---|
Max. Operating Temperature (Metric) | 80°C |
Display Type | None |
Voltage | 5-24 VDC |
Type | Flow Sensor |
Product Code | Brand | Waterflow Rate | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Waterflow Rate | Description | Price | Quantity & Availability | ||||
16649872
|
Masterflex™
24507-09 |
0.03 to 2 LPM | PVDF/Clamp Mount |
457.00 €
Each |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
16689872
|
Masterflex™
24507-32 |
0.03 to 2 LPM | PVDF/Clip mount |
277.00 €
Each |
Estimated Shipment: 20-03-2025
Log in to see stock. |
||||
16679862
|
Masterflex™
24507-02 |
0.03 to 2 LPM | PVDF/Tubeholder |
580.00 €
Each |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
16629872
|
Masterflex™
MFLX24507-07 |
0.06 to 2 LPM | PFA/Clamp Mount |
449.00 €
Each |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
16609872
|
Masterflex™
24507-05 |
0.06 to 2 LPM | PFA/Click Mount |
280.00 €
Each |
Estimated Shipment: 02-04-2025
Log in to see stock. |
||||
16669872
|
Masterflex™
24507-30 |
0.06 to 2 LPM | PFA/Clip Mount |
281.00 €
Each |
Estimated Shipment: 02-04-2025
Log in to see stock. |
||||
16639872
|
Masterflex™
MFLX24507-08 |
0.50 to 20 LPM | PFA/Clamp Mount |
470.00 €
Each |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
16619872
|
Masterflex™
24507-06 |
0.50 to 20 LPM | PFA/Click Mount |
291.00 €
Each |
Estimated Shipment: 02-04-2025
Log in to see stock. |
||||
16679872
|
Masterflex™
24507-31 |
0.50 to 20 LPM | PFA/Clip Mount |
288.00 €
Each |
Estimated Shipment: 02-04-2025
Log in to see stock. |
||||
16659872
|
Masterflex™
24507-10 |
0.50 to 20 LPM | PVDF/Clamp Mount |
477.00 €
Each |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
16699872
|
Masterflex™
MFLX24507-33 |
0.50 to 20 LPM | PVDF/Clip Mount |
274.00 €
Each |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
16689862
|
Masterflex™
24507-03 |
0.50 to 20 LPM | PVDF/Tubeholder |
600.00 €
Each |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
16669862
|
Masterflex™
24507-01 |
1.50 to 40 LPM | PFA/Clip Mount |
311.00 €
Each |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
16699862
|
Masterflex™
MFLX24507-04 |
3 to 200 LPM | PVDF/Tubeholder |
890.00 €
Each |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
Description
- Modular interchangeable flow sensor systems with single-use design
- Accurate measurement readings at low flow: ±1%
- Low operating cost, disposable flow tubes for optimal contamination-free production
- Ideal for single-use applications, but also suitable for long-term measurement
- Flow tubes and turbine sensors are produced and tested under cleanroom conditions
Specifications
176°F | |
None | |
Flow Sensor |
80°C | |
5-24 VDC |