Learn More
Siponimod, MedChemExpress
MedChemExpress Siponimod (BAF-312) is an orally active and selective sphingosine-1-phosphate (S1P) receptor modulator. Siponimod is selective for S1P1 and S1P5 over S1P2, S1P3, and S1P4, with EC50s of 0.4, 0.98, >10000, >1000, and 750 nM, respectively. Siponimod can be used for multiple sclerosis (MS) research-.
Brand: Medchem Express HY-12355
Description
Siponimod (BAF-312) is an orally active and selective sphingosine-1-phosphate (S1P) receptor modulator. Siponimod is selective for S1P1 and S1P5 over S1P2, S1P3, and S1P4, with EC50s of 0.4, 0.98, >10000, >1000, and 750 nM, respectively. Siponimod can be used for multiple sclerosis (MS) research-.Specifications
Siponimod | |
White | |
C29H35F3N2O3 | |
BAF-312 | |
O=C(C1CN(CC2=CC=C(/C(C)=N/OCC3=CC=C(C4CCCCC4)C(C(F)(F)F)=C3)C=C2CC)C1)O | |
516.6 | |
Research |
1230487-00-9 | |
Research | |
500 mg | |
DMSO : ≥ 30 mg/mL (58.07 mM) | |
516.6 | |
98.0% | |
Solid |