Learn More
Siponimod, MedChemExpress
MedChemExpress Siponimod (BAF-312) is an orally active and selective sphingosine-1-phosphate (S1P) receptor modulator. Siponimod is selective for S1P1 and S1P5 over S1P2, S1P3, and S1P4, with EC50s of 0.4, 0.98, >10000, >1000, and 750 nM, respectively. Siponimod can be used for multiple sclerosis (MS) research-.

91.80 € - 3068.55 €
Specifications
Chemical Name or Material | Siponimod |
---|---|
CAS | 1230487-00-9 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C29H35F3N2O3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16487837
![]() |
Medchem Express
HY-12355 |
5 mg |
91.80 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16471239
![]() |
Medchem Express
HY-12355 |
1 g |
3068.55 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16465118
![]() |
Medchem Express
HY-12355 |
10 mg |
137.70 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16431357
![]() |
Medchem Express
HY-12355 |
50 mg |
398.25 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16411636
![]() |
Medchem Express
HY-12355 |
500 mg |
1841.40 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16405846
![]() |
Medchem Express
HY-12355 |
100 mg |
552.15 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Siponimod (BAF-312) is an orally active and selective sphingosine-1-phosphate (S1P) receptor modulator. Siponimod is selective for S1P1 and S1P5 over S1P2, S1P3, and S1P4, with EC50s of 0.4, 0.98, >10000, >1000, and 750 nM, respectively. Siponimod can be used for multiple sclerosis (MS) research-.Specifications
Siponimod | |
White | |
C29H35F3N2O3 | |
DMSO : ≥ 30 mg/mL (58.07 mM) | |
516.6 | |
98.0% | |
Solid |
1230487-00-9 | |
Research | |
BAF-312 | |
O=C(C1CN(CC2=CC=C(/C(C)=N/OCC3=CC=C(C4CCCCC4)C(C(F)(F)F)=C3)C=C2CC)C1)O | |
516.6 | |
Research |
Safety and Handling
H302∣H315∣H319∣H335
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.