Learn More
Notrax Skystep™ ESD Rubber Mat with Moulded Edges
Ergonomically and economically engineered, the Skystep™ is designed to suction to the ground to prevent the mat from shifting while trapping air in its chambers. These air pockets combined with the resiliency of the rubber create an ultimate cushioning effect while keeping the mat lightweight.
Brand: Notrax 457S0034BL
Description
- Ergonomically and economically engineered, the Skystep™ utilizes the power of air
- Skystep™ is designed to suction to the ground to prevent the mat from shifting while trapping air in its chambers
- These air pockets combined with the resiliency of the rubber create an ultimate cushioning effect while keeping the mat lightweight
- Integrated bevelled edges on all four sides to prevent tripping on the mat
- Loose lay with no installation required
- Slip resistance rating R9 according to DIN 51130 and BG-rule BGR181
- Free of DOP, free of DMF, free of ozone-depleting substances, free of silicone and free of heavy metals
- Static dissipative natural rubber compound
- Meets IEC61340-4-1 (category DIF), measured resistance Rg 106 to 109 Ω, Rp 106 to 109 Ω
- Optional grounding cord
Specifications
Black | |
120 cm | |
Industrial and Commercial Matting |
47 in. | |
ESD Natural Rubber | |
90 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.