Learn More
Asecos™ SL-CLASSIC Safety Storage Cabinets
For the safe and proper storage of aggressive, non-flammable hazardous materials in working areas.
Brand:  Asecos™ 3061300130615
Product Code. 15600391
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Robust construction and longevity – tall cabinet with body made of powder-coated sheet steel and interior space made of melamine resin and flame retardant special plates
  • Safe and separate storage – acids and alkalis can be stored separately
  • Two hermetically divided storage compartments, integrated aeration and venting system, corrosion resistance of the metal-free air ducts
  • In accordance with TRGS 510
  • Adjusting aids are integrated in the cabinet base, fast and effective compensation for uneven floors
  • Interior equipment available includes pull-out shelves and bottom collecting sumps
  • No unauthorized use – doors lockable with profile cylinder (integration into an existing locking system possible)
  • Ventilation – integrated metal-free air ducts ready for connection to a technical exhaust system with only one extraction air inlet
Specifications
Specifications
TRGS 510
570 x 485 x 914 mm
Wing
300 kg
Powder-coated
Light Gray
Safety storage cabinet
1 x light gray wing door; 6 x PP pull-out shelves with PP sump, load capacity 25 kg, capacity 11 L
603 x 597 x 1965 mm
Storage of aggressive, non-flammable hazardous substances
1
176.4 kg
Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Asecos™ SL-CLASSIC Safety Storage Cabinets > 6x 25 kg/11 L PP pull-out shelves w/ sump