Learn More
Asecos™ SL-CLASSIC-UB Underbench Safety Storage Cabinets with 3 Doors
For the safe and proper storage of aggressive, non-flammable hazardous materials in working areas underneath work benches and fume cupboards.
Brand:  Asecos™ 3061689230618
 View more versions of this product
Product Code. 15955770
  • 1585.00 € / Each
Explore more special offers
Description
Description
  • Robust construction and longevity – underbench cabinets completely made of melamine resin and flame retardant special plates
  • Safe and separate storage – acids and alkalis can be stored separately
  • Two hermetically divided storage compartments, integrated aeration and venting system, corrosion resistance of the metal-free air ducts
  • In accordance with TRGS 510
  • Adjusting aids are integrated in the cabinet base, fast and effective compensation for uneven floors
  • Interior equipment available includes pull-out shelves and bottom collecting sumps
  • No unauthorized use – doors lockable with profile cylinder (integration into an existing locking system possible)
  • Ventilation – integrated metal-free air ducts ready for connection to a technical exhaust system with only one extraction air inlet
Specifications
Specifications
TRGS 510
531 x 489 x 562 mm
Wing
4 x Pull-out shelves: 25 kg/11 L each, Maximum load: 100 kg
103.5 kg
Melamine Resin
4 x 25 kg/11 L PP pull-out shelves with PP sump
570 x 1400 x 600 mm
Storage of aggressive and non-flammable hazardous substances
3
Light Gray
Safety storage cabinet