Learn More
BD Difco™ Salmonella O Antisera and Quality Control Antigens
Brand: BD 226451
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
Salmonella species cause a variety of human diseases called salmonelloses. The range of disease is from mild self-limiting gastroenteritis to more severe forms, possibly with bacteremia or typhoid fever, which can be life threatening. Hence diagnosis and identification of correct serotype is essential. These antisera aid in detecting such serotypes and the quality control antigens check their performance.
- Lyophilized, polyclonal rabbit antisera
- Polyvalent, specific for somatic antigens
- Cross-reactions are possible because serogroups may share non-major group antigens
- When properly rehydrated and used as recommended, each vial contains sufficient reagent for 60 tests
- For in vitro diagnostic use
- containing approximately 0.2% sodium azide as a preservative
Specifications
For in vitro diagnostic use | |
Antiserum |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Vial Sal O Antiserum Poly F Lyophilized
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Sodium Azide