missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassettes, 3.5K MWCO

Remove salts and contaminants, and maximize sample recovery using this semi-permeable membrane with a 3,500-Dalton protein MW cut-off (MWCO) cassette that is easier to use than traditional dialysis tubing.

153.00 € - 233.00 €

Specifications

Content And Storage Store at room temperature
Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Cassette
MWCO 3.5 kDa
View More Specs

Products 7
Product Code Brand Sample Volume (Metric) Quantity Price Quantity & Availability  
Product Code Brand Sample Volume (Metric) Quantity Price Quantity & Availability  
11859410
View Documents
Thermo Scientific™
66333
0.5 mL 10 Cassettes
153.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
10455685
View Documents
Thermo Scientific™
66335
0.5 mL 1 Kit (10 Cassettes)
168.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10759784
View Documents
Thermo Scientific™
66330
3 mL 10 Cassettes
156.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
10751565
View Documents
Thermo Scientific™
66332
3 mL 1 Kit (10 Cassettes)
186.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10500814
View Documents
Thermo Scientific™
66110
12 mL 8 Cassettes
198.00 €
Pack of 8
Please to purchase this item. Need a web account? Register with us today!
10416144
View Documents
Thermo Scientific™
66107
12 mL 1 Kit (8 Cassettes)
233.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10501004
View Documents
Thermo Scientific™
66130
30 mL 6 Cassettes
157.00 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

Store at room temperature
Cellulose Membrane, Dialysis
3.5 kDa
Buffer Exchange, Protein
Cassette
Slide-A-Lyzer™
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassettes, 3.5K MWCO

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.