Learn More
Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassettes, 3.5K MWCO
Remove salts and contaminants, and maximize sample recovery using this semi-permeable membrane with a 3,500-Dalton protein MW cut-off (MWCO) cassette that is easier to use than traditional dialysis tubing.
Brand: Thermo Scientific™ 66130
145.35 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Slide-A-Lyzer Dialysis Cassettes (3.5K MWCO) facilitate simple and effective removal of salts and small contaminants from proteins and other macromolecules larger than 3,500 Daltons. They can be used for a wide range of applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration.
Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips, or caps are needed to seal the units and prevent leaking and sample loss during the dialysis procedure. Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.Features of Slide-A-Lyzer Dialysis Cassettes:
• Easy to use—no knots or clamps are needed; just inject sample into cassette and begin dialysis
• Fast dialysis—flat cassette chamber with two membranes provides high surface area-to-volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
• High recovery—rectangular cassette design maximizes recovery of entire sample volume from any one of the four corner injection ports
• Four cassette sizes—select the cassette that best suits your sample volume (0.5 to 30 mL)
• Color-coded—pink cassette frame specifies cassettes with the 3,500 MW pore size membrane
Membrane specifications:
• Composition: regenerated cellulose synthesized by the viscose method
• Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
• Hydration time required before use: 2 minutes
• Glycerol content: none
• Sulfur content: 0.1–0.15%
• Heavy metals content: trace
Volume ranges:
• 0.5 mL = 0.1 to 0.5 mL
• 3 mL = 0.5 to 3 mL
• 12 mL = 3 to 12 mL
• 30 mL = 12 to 30 mL

Specifications
Store at room temperature | |
Cellulose Membrane, Dialysis | |
3.5 kDa | |
6 Cassettes |
Buffer Exchange, Protein | |
Cassette | |
Slide-A-Lyzer™ | |
30 mL |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P00002482 | Certificate of Analysis | 15/10/2024 | 66130 |
P00002484 | Certificate of Analysis | 15/10/2024 | 66130 |
P00002407 | Certificate of Analysis | 09/03/2024 | 66130 |
P00002332 | Certificate of Analysis | 25/01/2024 | 66130 |
P00002331 | Certificate of Analysis | 25/01/2024 | 66130 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.