Learn More
Ibidi μ-Slide I 0.4 Collagen IV
A μ-Slide has one channel for applying defined shear stress.
Brand:  Ibidi 80172
Encompass
Product Code. 17226153
  • POA / Pack of 15
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • A μ-Slide with one channel for cell cultivation under flow and for applying defined shear stress
  • easy to connect with tubing and pump systems via female Luers
  • channel height: 400 μm
  • bottom material: #1.5 Polymer Coverslip
  • Collagen IV coated
  • sterilized
Specifications
Specifications
Microscope Slide
15
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Ibidi μ-Slide I 0.4 Collagen IV