Learn More
Ibidi μ-Slide III 3D Perfusion Uncoated
μ-Slide III 3D Perfusion Uncoated is a three channel μ-Slide used for the cultivation and perfusion of 3D cell structure.
Brand:  Ibidi 80371
Encompass
Product Code. 15561255
  • POA / Pack of 15
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • A three channel slide used for the cultivation and perfusion of cells or spheroids in 3D matrices
  • easy to connect with tubing and pump systems via female Luers
  • bottom material: #1.5 Polymer Coverslip
  • hydrophobic
  • sterilized
Specifications
Specifications
Microscope Slide
15
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Ibidi μ-Slide III 3D Perfusion Uncoated