Learn More
Thermo Scientific™ Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Maximize tube storage capacity per freezer with a racking solution for five inner-door, ultra-low temperature freezers.
Brand:  Thermo Scientific™ 920099
 View more versions of this product
Product Code. 10731310
  • 198.99 € / Each
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 5 shelf Series Freezers as well as well as previous generation Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • Warranty varies by country, please contact us for details
  • TRUSTED_SUSTAINABILITY
Specifications
Specifications
Box Racks - Sliding Drawer
Silver
Stainless Steel
13 cu. ft. capacity upright freezers
5.5 in.
9.5 in.
44.2 cm
3 in. Box Rack - Sliding Drawer
Boxes (3 in.)
9 Boxes/Rack
14 cm
24 cm
16.5 in.