Learn More
Thermo Scientific™ Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Maximize tube storage capacity per freezer with a racking solution for five inner-door, ultra-low temperature freezers.
159.80 € - 344.72 €
Product Code | Brand | For Use With (Equipment) | Holds | No. of Racks | Positions | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Holds | No. of Racks | Positions | Price | Quantity & Availability | ||
15140509
|
Thermo Scientific™
1950780 |
13 cu. ft. EXF240, HFU240B | Tubes | 4 | 96/Box |
273.36 €
Each |
Estimated Shipment: 14-04-2025
Log in to see stock. |
||
15150509
|
Thermo Scientific™
1950782 |
17.3 and 23 cu. ft. EXF320, EXF400, HFU320B, HFU400B | Matrix™ 200 μL, 2D internally threaded universal tubes | 4 (320 Series) 5 (400 Series) | 36 Boxes/Rack |
287.43 €
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||
15130509
|
Thermo Scientific™
1950784 |
28 cu. ft. EXF600, HFU600B | Tubes | 6 | 96/Box |
338.69 €
Each |
Estimated Shipment: 10-04-2025
Log in to see stock. |
||
15111045
|
Thermo Scientific™
1950786 |
14.9, 19.4, 24.1, 28.8 and 33.5 cu. ft. Revco UxF, HERAFreeze HFU-T | Matrix™ 200 μL, 2D internally threaded universal tubes | 3 (300 capacity) 4 (400 capacity) 5 (500 capacity) 6 (600 capacity) 7 (700 capacity) | 42 Boxes/Rack |
344.72 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
||
11359291
|
Thermo Scientific™
1950637 |
13 cu. ft. EXF240, HFU240B (5 shelf) | Microplates | 4 | 16 Plates/Rack |
206.03 €
Each |
In Stock
Log in to see stock. |
||
11711101
|
Thermo Scientific™
1950638 |
17.3 and 23 cu. ft. EXF320, EXF400, HFU320B, HFU400B (5 shelf) | Microplates | 4 (320 Series) 5 (400 Series) | 24 Plates/Rack |
234.17 €
Each |
In Stock
Log in to see stock. |
||
15140539
|
Thermo Scientific™
1950730 |
28 cu. ft. EXF600, HFU600B | Matrix 2D Tubes (500 μL) | 6 | 28 Boxes/Rack |
273.36 €
Each |
Estimated Shipment: 14-04-2025
Log in to see stock. |
||
11379291
|
Thermo Scientific™
1950639 |
14.9, 19.4, 24.1, 28.8 and 33.5 cu. ft. Revco UxF, HERAFreeze HFU-T | Microplates | 3 (300 capacity) 4 (400 capacity) 5 (500 capacity) 6 (600 capacity) 7 (700 capacity) | 28 Plates/Rack |
253.26 €
Each |
In Stock
Log in to see stock. |
||
15160509
|
Thermo Scientific™
1950792 |
13 cu. ft. EXF240, HFU240B | Tubes | 4 | 96/Box |
210.05 €
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||
15170509
|
Thermo Scientific™
1950794 |
17.3 and 23 cu. ft. EXF320, EXF400, HFU320B, HFU400B | Matrix 2D Tubes (1 mL) | 4 (320 Series) 5 (400 Series) | 18 Boxes/Rack |
241.20 €
Each |
Estimated Shipment: 10-04-2025
Log in to see stock. |
||
15180509
|
Thermo Scientific™
1950796 |
28 cu. ft. EXF600, HFU600B | Matrix 2D Tubes (1 mL) | 6 | 21 Boxes/Rack |
261.30 €
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||
15190509
|
Thermo Scientific™
1950798 |
14.9, 19.4, 24.1, 28.8 and 33.5 cu. ft. Revco UxF, HERAFreeze HFU-T | Matrix 2D Tubes (1 mL) | 3 (300 capacity) 4 (400 capacity) 5 (500 capacity) 6 (600 capacity) 7 (700 capacity) | 21 Boxes/Rack |
261.30 €
Each |
In Stock
Log in to see stock. |
||
15100519
|
Thermo Scientific™
1950800 |
13 cu. ft. EXF240, HFU240B | Tubes | 4 | 48/Box |
159.80 €
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||
15110519
|
Thermo Scientific™
1950802 |
17.3 and 23 cu. ft. EXF320, EXF400, HFU320B, HFU400B | Nunc™ 5 mL 2D tubes or Matrix™ 5 mL 2D Tubes | 4 (320 Series) 5 (400 Series) | 24 Boxes/Rack |
165.83 €
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||
15130519
|
Thermo Scientific™
1950816 |
28 cu. ft. EXF600, HFU600B | Tubes | 6 | 48/Box |
201.00 €
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||
15120519
|
Thermo Scientific™
1950818 |
14.9, 19.4, 24.1, 28.8 and 33.5 cu. ft. Revco UxF, HERAFreeze HFU-T | Nunc™ 5 mL 2D tubes or Matrix™ 5 mL 2D Tubes | 3 (300 capacity) 4 (400 capacity) 5 (500 capacity) 6 (600 capacity) 7 (700 capacity) | 28 Boxes/Rack |
181.91 €
Each |
In Stock
Log in to see stock. |
||
11369311
|
Thermo Scientific™
398321 |
13 cu. ft. capacity upright freezers | Microplates with Lids | 70 Plates/Rack |
233.16 €
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||
11389311
|
Thermo Scientific™
398323 |
17.3 and 23 cu. ft. capacity upright freezers | Microplates with Lids | 84 Plates/Rack |
234.17 €
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||
11399311
|
Thermo Scientific™
398324 |
13 cu. ft. capacity upright freezers | Boxes (2 in.) | 12 Boxes/Rack |
205.02 €
Each |
Estimated Shipment: 08-04-2025
Log in to see stock. |
|||
11309321
|
Thermo Scientific™
398325 |
13 cu. ft. capacity upright freezers | Boxes (3 in.) | 9 Boxes/Rack |
205.02 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
|||
11319321
|
Thermo Scientific™
398326 |
17.3 and 23 cu. ft. capacity upright freezers | Boxes (2 in.) | 16 Boxes/Rack |
233.16 €
Each |
In Stock
Log in to see stock. |
|||
11359321
|
Thermo Scientific™
398330 |
17.3 and 23 cu. ft. capacity upright freezers | Boxes (3 in.) | 12 Boxes/Rack |
223.11 €
Each |
In Stock
Log in to see stock. |
|||
11369321
|
Thermo Scientific™
398331 |
17.3 and 23 cu. ft. capacity upright freezers | Boxes (2 in.) | 16 Boxes/Rack |
223.11 €
Each |
In Stock
Log in to see stock. |
|||
11379321
|
Thermo Scientific™
398332 |
13 cu. ft. capacity upright freezers | Boxes (2 in.) | 12 Boxes/Rack |
193.97 €
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||
11389321
|
Thermo Scientific™
398333 |
17.3, 23 and 28.8 cu. ft. capacity upright freezers | Microplates | 66 Plates/Rack |
233.16 €
Each |
Estimated Shipment: 16-04-2025
Log in to see stock. |
|||
11399321
|
Thermo Scientific™
398334 |
13 cu. ft. capacity upright freezers | Microplates | 44 Plates/Rack |
211.05 €
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||
10731310
|
Thermo Scientific™
920099 |
13 cu. ft. capacity upright freezers | Boxes (3 in.) | 9 Boxes/Rack |
198.99 €
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||
11376311
|
Thermo Scientific™
398335 |
17.3, 23 and 28.8 cu. ft. capacity upright freezers | Microplates | 30 Plates/Rack |
233.16 €
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||
11329321
|
Thermo Scientific™
398327 |
17.3 and 23 cu. ft. capacity upright freezers | Boxes (2 in.) | 12 Boxes/Rack |
233.16 €
Each |
In Stock
Log in to see stock. |
|||
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 5 shelf Series Freezers as well as well as previous generation Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
Thermo Scientific™ freezer racking and tubing solutions offer a convenient, streamlined process to maximize sample storage in your laboratory. Our racking solutions are designed to hold tubes ranging from 200μL up to 5.0mL, offering maximized sample storage capacity.
- With sliding drawer racks
- Adjustable side access box racks
- Standard/deepwell microplate racks
- Warranty varies by country, please contact us for details