Learn More
Thermo Scientific™ Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Maximize tube storage capacity per freezer with a racking solution for five inner-door, ultra-low temperature freezers.
Brand:  Thermo Scientific™ 398326
 View more versions of this product
Product Code. 11319321
  • 233.16 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 5 shelf Series Freezers as well as well as previous generation Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • Warranty varies by country, please contact us for details
  • TRUSTED_SUSTAINABILITY
Specifications
Specifications
Side Access Freezer Rack (Adjustable)
Silver
Stainless Steel
4 Tiers
13.7 cm
23.9 cm
22.1 in.
2 in. Box Racks - Adjustable side access rack
Boxes (2 in.)
16 Boxes/Rack
17.3 and 23 cu. ft. capacity upright freezers
5.4 in.
9.4 in.
56.1 cm