missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Slice™ Small Scissors
Small safety scissors cut tough materials like Kevlar-reinforced fabric
47.00 € - 71.00 €
Specifications
Material | GFN, TPE, Stainless Steel, Zirconium Oxide |
---|---|
Size | Small |
Color | White/Orange |
For Use With (Application) | Safety Cutting |
Includes | Zirconium oxide safety blades |
Description
Specifications
GFN, TPE, Stainless Steel, Zirconium Oxide | |
White/Orange | |
Zirconium oxide safety blades |
Small | |
Safety Cutting | |
1 Ea. |