missing translation for 'onlineSavingsMsg'
Learn More
3M™ Blue Disposable Coveralls, 4500 Series Product Code.: 13413686
Add to basket

3M™ Blue Disposable Coveralls, 4500 Series

Product Code. 13413686
X-Large, 70 to 74 in., 179 to 187 cm
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Height (English):
64 to 67 in.
66 to 69 in.
69 to 71 in.
70 to 74 in.
73 to 76 in.
76 to 78 in.
78 to 81 in.
Height (Metric):
164 to 170 cm
167 to 176 cm
174 to 181 cm
179 to 187 cm
186 to 194 cm
194 to 200 cm
200 to 206 cm
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 13413686

Brand: 3M™ 7000089600

Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Use for non-hazardous applications. The Blue Disposable Coveralls, 4500 Series offers Category I protection from polypropylene fabric.

CE Simple, Category 1 coverall is designed for domestic DIY use, light-duty industrial cleaning and maintenance applications, or environments where hygiene and cleanliness are important.
  • Excellent economic solution to keep the wearer clean and dry
  • Suitable for non-hazardous applications
  • Breathable and light-duty protection
  • Strong, durable 45gsm polypropylene fabric
  • Storm flap over zip
  • Elasticated hood, cuffs, ankles, and waist
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size X-Large
Gender Unisex
Material Polyethylene
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Hood Type 2-panel
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Protection Class Category I
Model 4500B
Size (English) Chest 43 to 45 in.
Size (Metric) Chest 108 to 115 cm
For Use With (Application) Light Industrial Cleaning, Maintenance
Height (English) 70 to 74 in.
Height (Metric) 179 to 187 cm
Show More Show Less
  • 3M Safety Solutions - Protective coveralls overview
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      3M Safety Solutions - Protective coveralls overview

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    3M™ Blue Disposable Coveralls, 4500 Series >

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.