missing translation for 'onlineSavingsMsg'
Learn More

Motic™ SMZ 160 Stereo Microscopes

This stereo zoom microscope comes with a 1:6.4 zoom range and an all metal body with classic illumination.

Brand:  Motic™ 1100201300161

 View more versions of this product

Product Code. 17804501

  • 774.00 € / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The SMZ-160 is ideal for educational purposes in high schools and universities, but is also great for industrial quality control.

  • Greenough Zoom Optical System
  • 45° stereomicroscope head
  • 6:1 zoom ratio
  • Diopter Adjustment: ±5
Specifications

Specifications

Plain Stand
Auxiliary Objectives (0.3X, 0.5X, 0.63X, 0.75X, 1.5X, 2X)
WF 10X/20 mm
10X
Trinocular
100 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.