missing translation for 'onlineSavingsMsg'
Learn More

Motic™ SMZ 160 Stereo Microscopes

This stereo zoom microscope comes with a 1:6.4 zoom range and an all metal body with classic illumination.

Brand:  Motic™ 1100201300151

 View more versions of this product

Product Code. 17847285

  • 656.00 € / Each
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The SMZ-160 is ideal for educational purposes in high schools and universities, but is also great for industrial quality control.

  • Greenough Zoom Optical System
  • 45° stereomicroscope head
  • 6:1 zoom ratio
  • Diopter Adjustment: ±5
Specifications

Specifications

Plain Stand
Auxiliary Objectives (0.3X, 0.5X, 0.63X, 0.75X, 1.5X, 2X)
WF 10X/20 mm
10X
Binocular
100 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers