missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium acetate, ReagentPlus™, Honeywell™
Reagent Grade, anhydrous,≥99.0%
30.73 € - 338.10 €
Chemical Identifiers
CAS | 127-09-3 |
---|---|
Molecular Formula | C2H3NaO2 |
Molecular Weight (g/mol) | 82.03 |
MDL Number | MFCD00012459 |
InChI Key | VMHLLURERBWHNL-UHFFFAOYSA-M |
Synonym | sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech |
PubChem CID | 517045 |
ChEBI | CHEBI:32954 |
SMILES | [Na+].CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15678120
|
Honeywell Research Chemicals
S8750-250G |
250 g | N/A | ||||||
15688120
|
Honeywell Research Chemicals
S8750-500G |
500 g | N/A | ||||||
15668120
|
Honeywell Research Chemicals
S8750-1KG |
1 kg | N/A | ||||||
15698120
|
Honeywell Research Chemicals
S8750-5KG |
5 kg | N/A | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
127-09-3 | |
82.03 | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
517045 | |
[Na+].CC([O-])=O |
C2H3NaO2 | |
MFCD00012459 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
CHEBI:32954 |
Specifications
127-09-3 | |
CH3COONa | |
NONH for all modes of transport | |
<250°C (>482°F) | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
sodium acetate | |
517045 | |
82.03g/mol | |
ReagentPlus™ | |
Sodium acetate |
C2H3NaO2 | |
MFCD00012459 | |
3595639 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
[Na+].CC([O-])=O | |
82.03 | |
CHEBI:32954 | |
≥99% | |
1112°F |
Safety and Handling
missing translation for 'rtecsNumber' : AJ4300010