missing translation for 'onlineSavingsMsg'
Learn More

Sodium azide, 99%, extra pure, Thermo Scientific Chemicals

24.42 € - 593.96 €

Chemical Identifiers

CAS 26628-22-8
MDL Number MFCD00003536
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10338380
SDS View Documents
Thermo Scientific Acros
190380050
5 g
24.42 €
5g
Please to purchase this item. Need a web account? Register with us today!
10592211
SDS View Documents
Thermo Scientific Acros
190381000
100 g
48.66 €
100g
Please to purchase this item. Need a web account? Register with us today!
10430471
SDS View Documents
Thermo Scientific Acros
190385000
500 g
186.05 €
500g
Please to purchase this item. Need a web account? Register with us today!
10124813
SDS View Documents
Thermo Scientific Acros
190380025
2.5 kg
593.96 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, For Azidation/Diazo transfer
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium azide, 99%, extra pure, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.