missing translation for 'onlineSavingsMsg'
Learn More

Sodium bicarbonate, 1M buffer soln., pH 8.5, Thermo Scientific Chemicals

73.67 € - 107.54 €

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15473179
View Documents Promotion Details
Thermo Scientific Alfa Aesar
J60408.AK
250 mL
73.67 €
250mL
Please to purchase this item. Need a web account? Register with us today!
15483179
View Documents Promotion Details
Thermo Scientific Alfa Aesar
J60408.AP
500 mL
107.54 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sodium bicarbonate acts a weak buffer with that makes maintaining a pH range of 7.2 - 7.6. It is also used as a nutrient source. This solution prepared in water is used as injection in Pharmaceutical industry.is used to treat cement contamination ins hore pac drilling slurry and bentonite water based drilling fluids.It is also used as a buffer. Sodium bicarbonate will remove calcium and manganese making hard water soft.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bicarbonate acts a weak buffer with that makes maintaining a pH range of 7.2 - 7.6. It is also used as a nutrient source. This solution prepared in water is used as injection in Pharmaceutical industry.is used to treat cement contamination ins hore pac drilling slurry and bentonite water based drilling fluids.It is also used as a buffer. Sodium bicarbonate will remove calcium and manganese making hard water soft.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong acids, strong oxidizing agents.
Specifications

Chemical Identifiers

144-55-8
84.01
UIIMBOGNXHQVGW-UHFFFAOYSA-M
CHEBI:32139
[Na+].OC([O-])=O
CHNaO3
MFCD00003528
516892
sodium;hydrogen carbonate

Specifications

144-55-8
8.5
CHNaO3
Soluble in water
[Na+].OC([O-])=O
84.01
CHEBI:32139
1 M
Liquid
MFCD00003528
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium;hydrogen carbonate
516892
Sodium bicarbonate buffer soln.
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only