Learn More
Sodium bismuthate(V), 85%, Thermo Scientific Chemicals
29.35 € - 312.56 €
Chemical Identifiers
CAS | 12232-99-4 |
---|---|
Molecular Formula | BiNaO3 |
Molecular Weight (g/mol) | 279.96 |
InChI Key | PNYYBUOBTVHFDN-UHFFFAOYSA-N |
Synonym | sodium bismuthate, bismuth sodium oxide, sodium bismuthate v, bismuth sodium trioxide, bismuthate bio31-, sodium, sodium oxido dioxo bismuth, sodium bismuth oxide, bio3.na, bismuthate bio31#-, sodium |
PubChem CID | 4063671 |
IUPAC Name | sodium;oxido(dioxo)bismuth |
SMILES | [O-][Bi](=O)=O.[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10012770
|
Thermo Scientific Acros
205210250 |
25 g | Glass bottle |
29.35 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10195020
|
Thermo Scientific Acros
205212500 |
250 g | Glass bottle |
84.92 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11904341
|
Thermo Scientific Acros
205210010 |
1 kg | Glass bottle |
312.56 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12232-99-4 | |
279.96 | |
sodium bismuthate, bismuth sodium oxide, sodium bismuthate v, bismuth sodium trioxide, bismuthate bio31-, sodium, sodium oxido dioxo bismuth, sodium bismuth oxide, bio3.na, bismuthate bio31#-, sodium | |
sodium;oxido(dioxo)bismuth |
BiNaO3 | |
PNYYBUOBTVHFDN-UHFFFAOYSA-N | |
4063671 | |
[O-][Bi](=O)=O.[Na+] |
Specifications
12232-99-4 | |
Powder | |
BiNaO3 | |
15,8721 | |
Solubility in water: insoluble. Other solubilities: decomposes in hot water and acids | |
[O-][Bi](=O)=O.[Na+] | |
279.96 | |
279.96 | |
Sodium bismuthate(V) |
Yellow to Brown | |
80% min. (Iodometry) | |
NaBiO3 | |
sodium bismuthate, bismuth sodium oxide, sodium bismuthate v, bismuth sodium trioxide, bismuthate bio31-, sodium, sodium oxido dioxo bismuth, sodium bismuth oxide, bio3.na, bismuthate bio31#-, sodium | |
PNYYBUOBTVHFDN-UHFFFAOYSA-N | |
sodium;oxido(dioxo)bismuth | |
4063671 | |
85% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
If skin irritation occurs: Get medical advice/a
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 235-455-6
- Sodium bismuthate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Bismuth sodium oxide
RUO – Research Use Only