missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium chloride, Puriss., meets analytical spec. of BP, Ph. Eur., USP, 99.0-100.5% (calc. to the dried substance), ≤0.00002% Al, Honeywell Fluka™
Puriss., meets analytical specification of BP, Ph. Eur., USP, 99.0-100.5% (calc. to the dried substance), ≤0.00002% Al
55.28 € - 683.09 €
Chemical Identifiers
CAS | 7647-14-5 |
---|---|
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
SMILES | [Na+].[Cl-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15681300
|
Honeywell-Fluka
13423-1KG |
1 kg |
55.28 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15621310
|
Honeywell-Fluka
13423-5KG |
5 kg |
163.23 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15631310
|
Honeywell-Fluka
13423-6X1KG |
6 x 1 kg |
261.33 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15671300
|
Honeywell-Fluka
13423-10KG |
10 kg | |||||||
15611310
|
Honeywell-Fluka
13423-4X5KG |
4 x 5 kg |
569.82 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15691300
|
Honeywell-Fluka
13423-25KG |
25 kg |
531.09 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15601310
|
Honeywell-Fluka
13423-32KG |
32 kg | |||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7647-14-5 | |
58.44 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
5234 | |
[Na+].[Cl-] |
ClNa | |
MFCD00003477 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
CHEBI:26710 |
Specifications
7647-14-5 | |
NaCl | |
NONH for all modes of transport | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
99 to 100.5% (calc. to the dried substance) | |
Sodium Chloride, Meets analytical specification of Ph. Eur.,BP, USP |
ClNa | |
MFCD00003477 | |
3534976 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium chloride | |
5234 | |
58.44g/mol | |
puriss. |
Safety and Handling
missing translation for 'einecsNumber' : 231-598-3
missing translation for 'rtecsNumber' : VZ4725000