missing translation for 'onlineSavingsMsg'
Learn More
Learn More

- Home
- Products
- Chemicals
- Salts and Inorganics
- Inorganic Salts
- Alkali Metal Salts
- Sodium chloride, ReagentPlus™, Honeywell™
Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
Sodium chloride, ReagentPlus™, Honeywell™
Product Code. 156681405 kg
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg
This item is not returnable.
View return policy
Product Code. 15668140
Brand: Honeywell Research Chemicals S96255KG
This item is not returnable.
View return policy
Description
- ≥95% for general laboratory use.

Specifications
Chemical Identifiers
CAS | 7647-14-5 |
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
SMILES | [Na+].[Cl-] |
Specifications
Quantity | 5 kg |
Linear Formula | NaCl |
UN Number | NONH for all modes of transport |
Beilstein | 3534976 |
IUPAC Name | sodium chloride |
Formula Weight | 58.44g/mol |
Percent Purity | ≥99% |
Grade | ReagentPlus™ |
Chemical Name or Material | Sodium chloride |
Product Suggestions
You may be interested in these alternatives
-
-
-
-
-
-
Product Code. 10216410
Customers who viewed this item also viewed This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
Viewing 1-5 of 6
-
Catalog No. 10703811
382,00 € / 500g -
Catalog No. 10783611
61,60 € / 500g -
Catalog No. 10117113
34,57 € / 100g -
Catalog No. 10112640
33,95 € / 1kg -
Catalog No. 10092740
27,80 € / 1kg -
Catalog No. 10401841
27,10 € / 500mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium chloride, ReagentPlus™, Honeywell™ >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Spot an opportunity for improvement?Share a Content Correction