Learn More
Sodium chromate tetrahydrate, 99+%, Thermo Scientific Chemicals
Description
Sodium chromate tetrahydrate is used as a corrosion inhibitor, wood preservative and an auxiliary in textile industry. It serves as an oxidant, which is used to prepare carboxylic acids and ketones from primary and secondary alcohols respectively. Further, it is used as a starting material to prepare other chromium compounds and as a catalyst in organic synthesis especially pharmaceuticals and agro chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium chromate tetrahydrate is used as a corrosion inhibitor, wood preservative and an auxiliary in textile industry. It serves as an oxidant, which is used to prepare carboxylic acids and ketones from primary and secondary alcohols respectively. Further, it is used as a starting material to prepare other chromium compounds and as a catalyst in organic synthesis especially pharmaceuticals and agro chemicals.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible withstrong reducing agents, organic materials, strong acids, combustible materials and powdered metals.

Specifications
Specifications
Quantity | 100 g |
UN Number | UN3288 |
Merck Index | 14,8601 |
Solubility Information | Soluble in water. |
Formula Weight | 234.04 (161.97 Anhydrous) |
Percent Purity | ≥99% |
Odor | Odorless |
Density | 2.73 |
Chemical Name or Material | Sodium chromate tetrahydrate |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10253323 | Certificate of Analysis | 23/12/2024 | A17499.22,A17499.36 |
10251767 | Certificate of Analysis | 30/08/2024 | A17499.22,A17499.36 |
10244808 | Certificate of Analysis | 25/11/2022 | A17499.22,A17499.36 |
10244208 | Certificate of Analysis | 19/10/2022 | A17499.22,A17499.36 |
10150951 | Certificate of Analysis | 23/11/2021 | A17499.22,A17499.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.