Learn More
Sodium cyanoborohydride, 95%, Thermo Scientific Chemicals
51.96 € - 611.04 €
Chemical Identifiers
CAS | 25895-60-7 |
---|---|
Molecular Formula | CH3BNNa |
Molecular Weight (g/mol) | 62.84 |
MDL Number | MFCD00003516 |
InChI Key | CVDUGUOQTVTBJH-UHFFFAOYSA-N |
Synonym | sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1- |
PubChem CID | 20587905 |
SMILES | [Na+].[BH3-]C#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10541231
|
Thermo Scientific Acros
168550100 |
10 g | Glass bottle |
51.96 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10082110
|
Thermo Scientific Acros
168550500 |
50 g | Glass bottle |
170.85 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10703504
|
Thermo Scientific Acros
168552500 |
250 g | Glass bottle |
611.04 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemische Identifikatoren
25895-60-7 | |
62.84 | |
CVDUGUOQTVTBJH-UHFFFAOYSA-N | |
20587905 |
CH3BNNa | |
MFCD00003516 | |
sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1-Show More | |
[Na+].[BH3-]C#N |
Spezifikation
>242.0°C | |
White | |
95% | |
NaBH3CN | |
04,448; 05,607; 06,537; 07,334; 08,454; 09,424; 10,360; 11,481; 12,445; 14,287; 16,305; 17,388 | |
sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1- | |
CVDUGUOQTVTBJH-UHFFFAOYSA-N | |
sodium;cyanoboron(1-) | |
20587905 | |
95% |
25895-60-7 | |
Powder | |
CH3BNNa | |
MFCD00003516 | |
15, 8742 | |
Solubility in water: 2120g/L at 29°C (slow decomposition). Other solubilities: soluble in methanol; soluble in thf, 372g/L, at 28°C, 410g/L at 46°C, 422g/L at 62°C, soluble in diglyme 176g/L at 25°C, slightly soluble in ethanol, insoluble in ether, b | |
[Na+].[BH3-]C#N | |
62.84 | |
62.84 | |
Sodium cyanoborohydride |
Sicherheit und Handhabung
GHS H Statement
Flammable solid.
In contact with water releases flammable gas.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic lif
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immedia
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 247-317-2
Recommended Storage : the substance is sensitive to gradually degrading on storage.
- Sodium cyanoborohydride
Signalwort
- Danger
Gefahrenkategorie
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 1 B
- Substance which\, in contact with water\, emits flammable gases Category 2
Gefahrenhinweis
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H261-In contact with water releases flammable gases.
- H228-Flammable solid.
- EUH032-Contact with acids liberates very toxic gas.
Sicherheitshinweis
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.