missing translation for 'onlineSavingsMsg'
Learn More

Sodium formate, ACS reagent, Thermo Scientific Chemicals

21.11 € - 194.97 €

Chemical Identifiers

CAS 141-53-7
Molecular Formula CHNaO2
Molecular Weight (g/mol) 68.01
InChI Key HLBBKKJFGFRGMU-UHFFFAOYSA-M
Synonym sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
PubChem CID 2723810
ChEBI CHEBI:62965
IUPAC Name sodium;formate
SMILES C(=O)[O-].[Na+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
12386263
View Documents Promotion Details
Thermo Scientific Acros
446951000
100 g Plastic bottle
21.11 €
100g
Please to purchase this item. Need a web account? Register with us today!
12357183
View Documents Promotion Details
Thermo Scientific Acros
446955000
500 g Plastic bottle
60.70 €
500g
Please to purchase this item. Need a web account? Register with us today!
12347183
View Documents Promotion Details
Thermo Scientific Acros
446950025
2.5 kg Plastic bottle
194.97 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

141-53-7
68.01
sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
CHEBI:62965
C(=O)[O-].[Na+]
CHNaO2
HLBBKKJFGFRGMU-UHFFFAOYSA-M
2723810
sodium;formate

Specifications

141-53-7
360.0°C
HCO2Na
sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
HLBBKKJFGFRGMU-UHFFFAOYSA-M
sodium;formate
2723810
68.01
0.005% max
258.0°C to 261.0°C
CHNaO2
158756
Solubility in water: 550g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol,insoluble in ether
C(=O)[O-].[Na+]
68.01
CHEBI:62965
ACS Reagent
Sodium formate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 205-488-

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only