missing translation for 'onlineSavingsMsg'
Learn More
Sodium hydride, 60% dispersion in mineral oil, in soluble bags, in resealable cans, Thermo Scientific Chemicals Product Code.: 10297520

Sodium hydride, 60% dispersion in mineral oil, in soluble bags, in resealable cans, Thermo Scientific Chemicals

Product Code. 10297520
100 g
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10297520

Brand: Thermo Scientific Acros 332141000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-69-7, 8042-47-5
Molecular Formula HNa
Molecular Weight (g/mol) 24
MDL Number MFCD00003471
InChI Key BZKBCQXYZZXSCO-UHFFFAOYSA-N
Synonym sodium hydride, sodiumhydride, sodium hydride nah, sodium hydride oil dispersion, sodium monohydride, sodium hydride nad, sodiumhydrid, natrium hydride, sodium hydridShow MoreShow Less
PubChem CID 24758
IUPAC Name sodium;hydride
SMILES [H-].[Na+]

Specifications

Melting Point 800.0°C
Color Gray to Yellow
Physical Form Tacky Powder
Quantity 100 g
Linear Formula NaH
Fieser 02,382; 01,1075; 04,452; 05,610; 06,541; 07,335; 08,458; 09,427; 11,486; 12,447; 14,288; 16,307
Flash Point 165°C
Merck Index 15,876
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohols, soluble in molten sodium hydroxide, insoluble in cs2, ccl4, liquid nh3, benzene
Formula Weight 24
Percent Purity 57 to 63% (NaH)
Packaging Plastic bag in resealable tin can
Density 1.2000g/mL
Chemical Name or Material Sodium hydride, 60% dispersion in mineral oil, In soluble bags, in resealable cans
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0402180 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number332141000,332140020,332145000
Lot NumberA0395478 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number332141000,332140020,332145000
Lot NumberA0403795 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number332141000,332140020,332145000
Lot NumberA0396618 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number332141000,332140020,332145000
Lot NumberA0395477 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number332141000,332140020,332145000
compliance-icons
Product Identifier
  • Sodium hydride, 60% dispersion in mineral oil
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium hydride, 60% dispersion in mineral oil, in soluble bags, in resealable cans, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.