missing translation for 'onlineSavingsMsg'
Learn More
Sodium Hydroxide 1.0M, Low in Carbonate, Analytical Volumetric Solution, Reagecon™ Product Code.: 11512992

Sodium Hydroxide 1.0M, Low in Carbonate, Analytical Volumetric Solution, Reagecon™
BIOPHARMA

Product Code. 11512992 Shop All Reagecon Products
1 L
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy

Product Code. 11512992

Brand: Reagecon S21001LC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides a convenient method for accurate volumetric analysis in the laboratory. Reagecon Sodium Hydroxide 1.0M, Low in Carbonate, Analytical Volumetric Solution is a cost-effective answer to time- and labor-consuming preparation problems.

Ready to use aqueous volumetric solution for all analytical laboratory applications, which can be used for manual or automatic titrations. This product is certified to a specification as outlined in the Certificate of Analysis. Lot Number and Expiry Date are printed on the label. Solution is certified traceable to N.I.S.T and this will be stated in the Certificate of Analysis.

  • Molecular Formula: NaOH
  • Molar Mass: 40.00g/mol
  • CAS: 1310-73-2
  • EIN ECS No.: 215-185-5
  • Low in carbonate (CO3)

Specifications

Color Colorless
Boiling Point 100°C
CAS 1310-73-2
Melting Point 0°C
Molecular Formula HNaO
Concentration 1.0mol/L
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Fully miscible
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40.00g/mol
Traceability to NIST Traceable to NIST
Vapor Pressure 23hPa (at 20°C)
Physical Form Liquid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS2124L1 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberS21001LC
Lot NumberS2124D1 Certificate TypeCertificate of Analysis Date17/12/2024 Catalog NumberS21001LC
Lot NumberS2123M1 Certificate TypeCertificate of Analysis Date24/04/2024 Catalog NumberS21001LC
Lot NumberS2123E1 Certificate TypeCertificate of Analysis Date16/11/2023 Catalog NumberS21001LC
Lot NumberS2122F1 Certificate TypeCertificate of Analysis Date07/03/2023 Catalog NumberS21001LC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Hydroxide 1.0M, Low in Carbonate, Analytical Volumetric Solution, Reagecon™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.