missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide 1.0M, Analytical Volumetric Solution, Reagecon™
Alle ansehen Reagecon Produkte

missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
Packaging:
1L
Beschreibung
This product is certified to a specification as outlined in the Certificate of Analysis. Lot Number and Expiry Date are printed on the label. Solution is certified traceable to N.I.S.T and this will be stated in the Certificate of Analysis.
- Molecular Formula: NaOH
- Molar Mass: 40.00g/mol
- CAS: 1310-73-2
- EIN ECS No.: 215-185-5
Spezifikation
Spezifikation
Color | Colorless |
Boiling Point | 100°C |
CAS | 1310-73-2 |
Melting Point | 0°C |
Molecular Formula | HNaO |
Concentration | 1.0M |
Density | 1.04387g/cm3 |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
Solubility Information | Fully miscible |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Mehr anzeigen |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Sicherheit und Handhabung

Produktidentifikator
- SODIUM HYDROXIDE 1.0N (1.0M)
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H314-Causes severe skin burns and eye damage.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : sodium hydroxide
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Sodium Hydroxide 1.0M, Analytical Volumetric Solution, Reagecon™ >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur