missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, ≥98% (Acidimetric), BioXtra, Honeywell
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
Description
- Stringent testing protocols may include trace metal analysis and specialized suitability testing, such as hybridoma or mouse embryo
- Includes application-specific formulations, such as tablets and prepared solutions.

Specifications
Specifications
Linear Formula | NaOH |
UN Number | UN1823 |
Quantity | 1 kg |
Formula Weight | 40g/mol |
Percent Purity | ≥98% (acidimetric) |
Vapor Density | >1 (vs air) |
Vapor Pressure | 18 mmHg (20°C), 3 mmHg (37°C) |
Chemical Name or Material | Sodium hydroxide |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Safety and Handling

Product Identifier
- Sodium hydroxide
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Hydroxide, ≥98% (Acidimetric), BioXtra, Honeywell >
Spot an opportunity for improvement?Share a Content Correction