missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide solution, Purum, ≥32%, Honeywell Fluka™
Purum,≥32%
70.60 € - 416.74 €
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
Molecular Weight (g/mol) | 39.997 |
MDL Number | MFCD00003548 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
IUPAC Name | sodium;hydroxide |
SMILES | [OH-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15670790
|
Honeywell-Fluka
05211-2.5L |
2.5 L | N/A | ||||||
15610800
|
Honeywell-Fluka
05211-5L |
5 L | N/A | ||||||
15690790
|
Honeywell-Fluka
05211-4X2.5L |
4 x 2.5 L | N/A | ||||||
15600800
|
Honeywell-Fluka
05211-4X5L |
4 x 5 L | N/A | ||||||
15680790
|
Honeywell-Fluka
05211-35KG |
35 kg | N/A | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Specifications
1310-73-2 | |
NaOH | |
1824 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40g/mol | |
purum | |
Sodium Hydroxide Solution |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
≥32% | |
3mmHg (37°C) |
Safety and Handling
P280-P305 + P351 + P338-P310
H290-H314
missing translation for 'einecsNumber' : 215-185-5