missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hypochlorite solution, purum, ≈10% (RT), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
2.5 L
10 L
Description
- ≥95% for general laboratory use.

Specifications
Specifications
Quantity | 2.5 L |
Linear Formula | NaClO |
UN Number | UN1791 |
Formula Weight | 74.44g/mol |
Percent Purity | 10% (RT) |
Grade | purum |
Concentration or Composition (by Analyte or Components) | ≈10% (RT) |
Density | 1.21-1.26g/mL at 20°C |
Chemical Name or Material | Sodium hypochlorite solution |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Safety and Handling


Product Identifier
- Sodium hypochlorite solution
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH031-Contact with acids liberates toxic gas.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : sodium hypochlorite,solution
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium hypochlorite solution, purum, ≈10% (RT), Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction