missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hypochlorite solution, purum, ≈10% (RT), Honeywell Fluka™
Purum, ∼10% (RT)
83.99 € - 274.27 €
Chemical Identifiers
CAS | 7681-52-9 |
---|---|
Molecular Formula | ClNaO |
Molecular Weight (g/mol) | 74.439 |
MDL Number | MFCD00011120 |
InChI Key | SUKJFIGYRHOWBL-UHFFFAOYSA-N |
Synonym | sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution |
PubChem CID | 23665760 |
ChEBI | CHEBI:32146 |
IUPAC Name | sodium;hypochlorite |
SMILES | [O-]Cl.[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15666830
|
Honeywell-Fluka
71696-2.5L |
2.5 L |
83.99 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15656830
|
Honeywell-Fluka
71696-10L |
10 L |
274.27 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7681-52-9 | |
74.439 | |
SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
23665760 | |
sodium;hypochlorite |
ClNaO | |
MFCD00011120 | |
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution | |
CHEBI:32146 | |
[O-]Cl.[Na+] |
Specifications
7681-52-9 | |
NaClO | |
UN1791 | |
SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
sodium;hypochlorite | |
23665760 | |
74.44g/mol | |
purum | |
1.21-1.26g/mL at 20°C |
ClNaO | |
MFCD00011120 | |
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution | |
[O-]Cl.[Na+] | |
74.439 | |
CHEBI:32146 | |
10% (RT) | |
≈10% (RT) | |
Sodium hypochlorite solution |
Safety and Handling
P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P391
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sodium hypochlorite solution
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : sodium hypochlorite,solution