missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Iodide, puriss., Honeywell Fluka™
Puriss., meets analytical specification of BP, Ph. Eur., USP, 99-100.5% (calc. to the dried substance)
44.21 € - 1470.54 €
Chemical Identifiers
CAS | 7681-82-5 |
---|---|
Molecular Formula | INa |
Molecular Weight (g/mol) | 149.894 |
MDL Number | MFCD00003532 |
InChI Key | FVAUCKIRQBBSSJ-UHFFFAOYSA-M |
Synonym | sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech |
PubChem CID | 5238 |
ChEBI | CHEBI:33167 |
IUPAC Name | sodium;iodide |
SMILES | [Na+].[I-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15670670
|
Honeywell-Fluka
03129-100G |
100 g | |||||||
15690670
|
Honeywell-Fluka
03129-500G |
500 g |
178.98 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15680670
|
Honeywell-Fluka
03129-1KG |
1 kg |
298.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15610680
|
Honeywell-Fluka
03129-6X500G |
6 x 500 g | |||||||
15600680
|
Honeywell-Fluka
03129-6X1KG |
6 x 1 kg |
1470.54 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7681-82-5 | |
149.894 | |
FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
5238 | |
sodium;iodide |
INa | |
MFCD00003532 | |
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech | |
CHEBI:33167 | |
[Na+].[I-] |
Specifications
7681-82-5 | |
NaI | |
3077 | |
FVAUCKIRQBBSSJ-UHFFFAOYSA-M | |
sodium;iodide | |
5238 | |
149.89g/mol | |
puriss. | |
Vapor Density: >1 (vs air) |
INa | |
MFCD00003532 | |
sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech | |
[Na+].[I-] | |
149.894 | |
CHEBI:33167 | |
99 to 100.5% (calc. to the dried substance) | |
Plastic Bottle | |
Sodium Iodide, Meets analytical specification of Ph.Eur. BP, USP |
Safety and Handling
P273-P305 + P351 + P338
H315-H319-H400
missing translation for 'einecsNumber' : 231-679-3
missing translation for 'rtecsNumber' : WB6475000