missing translation for 'onlineSavingsMsg'
Learn More

Sodium methoxide, pure, 5.4M (30 wt.%) solution in methanol, Thermo Scientific Chemicals

30.35 € - 462.30 €

Chemical Identifiers

CAS 124-41-4, 67-56-1
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.02
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10357680
SDS View Documents
Thermo Scientific Acros
168600250
25 mL
30.35 €
25mL
Please to purchase this item. Need a web account? Register with us today!
10347680
SDS View Documents
Thermo Scientific Acros
168600010
1 L
43.01 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10449090
SDS View Documents
Thermo Scientific Acros
168600025
2.5 L
75.78 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10357853
SDS View Documents
Thermo Scientific Acros
168600100
10 L
202.01 €
10 L
Please to purchase this item. Need a web account? Register with us today!
15294654
SDS View Documents
Thermo Scientific Acros
168600251
25 L
462.30 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

124-41-4, 67-56-1
54.02
WQDUMFSSJAZKTM-UHFFFAOYSA-N
10942334
C[O-].[Na+]
CH3NaO
MFCD00012179
sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
sodium;methanolate

Specifications

124-41-4, 67-56-1
Colorless to Yellow
93.0°C
Glass Bottle
CH3ONa
MFCD00012179
01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352
0.97
Solubility in water: reacts. Other solubilities: soluble in methanol
C[O-].[Na+]
54.02
68 mPa.s (20°C)
Pure
Sodium methoxide, 30 wt% Solution in Methanol
1.0°C to 5.0°C
0.9700g/mL
33°C
CH3NaO
1.3701
01,28
15,8775
sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
WQDUMFSSJAZKTM-UHFFFAOYSA-N
sodium;methanolate
10942334
54.02
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes damage to organs.
Flammable liquid and vapor.
May be corrosive to metals.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 204-699-5

Product Identifier
  • Sodium methoxide, 5.4M (30 wt.%) solution in methanol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Methanol, Sodium methylate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.