Learn More
Sodium methoxide, pure, anhydrous powder, Thermo Scientific Chemicals
24.32 € - 243.21 €
Chemical Identifiers
CAS | 124-41-4, 497-19-8, 1310-73-2 |
---|---|
Molecular Formula | CH3NaO |
Molecular Weight (g/mol) | 54.02 |
MDL Number | MFCD00012179 |
InChI Key | WQDUMFSSJAZKTM-UHFFFAOYSA-N |
Synonym | sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755Show More |
PubChem CID | 10942334 |
IUPAC Name | sodium;methanolate |
SMILES | C[O-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10327500
|
Thermo Scientific Acros
173120050 |
5 g | Glass bottle |
24.32 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10687953
|
Thermo Scientific Acros
173121000 |
100 g | Glass bottle |
36.48 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10675042
|
Thermo Scientific Acros
173125000 |
500 g | Glass bottle |
58.79 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10236873
|
Thermo Scientific Acros
173120025 |
2.5 kg | Plastic Drum |
243.21 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
124-41-4, 497-19-8, 1310-73-2 | |
54.02 | |
WQDUMFSSJAZKTM-UHFFFAOYSA-N | |
10942334 | |
C[O-].[Na+] |
CH3NaO | |
MFCD00012179 | |
sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755Show More | |
sodium;methanolate |
Specifications
total alkalinity (as NaOCH3):> 99% | |
126.0°C | |
0.8 | |
CH3ONa | |
1431 | |
01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352 | |
sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755 | |
WQDUMFSSJAZKTM-UHFFFAOYSA-N | |
sodium;methanolate | |
10942334 | |
≥97% (NaOCH3) | |
Powder |
124-41-4, 497-19-8, 1310-73-2 | |
White-Yellow | |
CH3NaO | |
MFCD00012179 | |
01, 280 | |
15, 8775 | |
Solubility in water: reacts with water. Other solubilities: soluble in methanol and ethanol, insoluble in hydrocarbons(benzene, toluene, xylene | |
C[O-].[Na+] | |
54.02 | |
54.02 | |
Pure | |
Sodium methoxide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Self-heating; may catch fire.
Flammable solid.
Harmful if swallowed.
May be corrosive to metals.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 204-699-5
missing translation for 'rtecsNumber' : PC3570000
missing translation for 'tsca' : TSCA
- Sodium methoxide, anhydrous powder
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 1
- Substances and mixtures corrosive to metals Category 1
- Self-heating substance Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H251-Self-heating: may catch fire.
- H290-May be corrosive to metals.
- H228-Flammable solid.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235+P410-Keep cool. Protect from sunlight.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.