missing translation for 'onlineSavingsMsg'
Learn More
Sodium nitrite, 0.1M Standardized Solution, Thermo Scientific Chemicals Product Code.: 15411857

Sodium nitrite, 0.1M Standardized Solution, Thermo Scientific Chemicals

Product Code. 15411857
500 mL
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy

Product Code. 15411857

Brand: Thermo Scientific Alfa Aesar 043015.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

Sodium nitrite, 0.1M standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7632-00-0, 7732-18-5
Quantity 500 mL
Vapor Pressure 17mm Hg at 20°C
Density 1.01g/cm3 at 20°C
Molecular Formula NNaO2
MDL Number MFCD00011118
Physical Form Liquid
Color Colorless
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
SMILES [Na+].[O-]N=O
IUPAC Name sodium;nitrite
Solubility Miscible with water, methanol, ethanol, ammonia and nitric acid; Slightly miscible with diethyl ether
Molecular Weight (g/mol) 69.00
PubChem CID 23668193
ChEBI CHEBI:78870
Formula Weight 69
Concentration or Composition (by Analyte or Components) Sodium nitrite: 0.7%; Water: 99.3%
Chemical Name or Material Sodium nitrite
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM04H034 Certificate TypeCertificate of Analysis Date20/02/2025 Catalog Number043015.AP
Lot NumberX24K002 Certificate TypeCertificate of Analysis Date08/11/2024 Catalog Number043015.AP
Lot NumberP27K026 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog Number043015.AP
Lot NumberP27K025 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog Number043015.AP
Lot NumberM17K014 Certificate TypeCertificate of Analysis Date23/01/2024 Catalog Number043015.AP

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium nitrite, 0.1M Standardized Solution, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.