missing translation for 'onlineSavingsMsg'
Learn More
Sodium nitroferricyanide dihydrate, 99+%, Thermo Scientific Chemicals Product Code.: 10561461

Sodium nitroferricyanide dihydrate, 99+%, Thermo Scientific Chemicals

Product Code. 10561461
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10561461

Brand: Thermo Scientific Acros 211640250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13755-38-9
Molecular Formula C5H4FeN6Na2O3
Molecular Weight (g/mol) 297.95
MDL Number MFCD00149192
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2oShow MoreShow Less
PubChem CID 11953895
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

Specifications

Color Red
Physical Form Crystals
Quantity 25 g
Assay Percent Range 99% min. (Argentometry)
Linear Formula Na2Fe(CN)5NO·2H2O
Merck Index 15, 8780
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: slightly soluble in alcohol
IUPAC Name disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate
Formula Weight 297.94
Percent Purity ≥99%
Packaging Glass bottle
Specific Gravity 1.72
Density 1.7200g/mL
Chemical Name or Material Sodium nitroferricyanide dihydrate, p.a.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465374 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog Number211645000,211640250,211641000
Lot NumberA0453956 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number211645000,211640250,211641000
Lot NumberA0409712 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number211645000,211640250,211641000
Lot NumberA0410666 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number211645000,211640250,211641000
Lot NumberA0407096 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number211645000,211640250,211641000
compliance-icons
Product Identifier
  • Sodium nitroferricyanide dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium nitroferricyanide dihydrate, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.