missing translation for 'onlineSavingsMsg'
Learn More
Sodium Nitroprusside Dihydrate, puriss. p.a., ≥99%, ACS reagent, Honeywell Fluka™ Product Code.: 15686840

Sodium Nitroprusside Dihydrate, puriss. p.a., ≥99%, ACS reagent, Honeywell Fluka™

Product Code. 15686840 Shop All Honeywell Fluka Products
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 15686840

Brand: Honeywell-Fluka 71778100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reagent European Pharmacopoeia, Reagent European Pharmacopoeia,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13755-38-9
Molecular Formula C5H4FeN6Na2O3
Molecular Weight (g/mol) 297.95
MDL Number MFCD00149192
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
PubChem CID 11953895
IUPAC Name disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

Specifications

Quantity 100 g
Linear Formula Na2[Fe(CN)5NO] · 2H2O
UN Number UN3288
Formula Weight 297.95g/mol
Percent Purity ≥99%
Chemical Name or Material Sodium nitroprusside dihydrate
compliance-icons
Product Identifier
  • Sodium nitroprusside dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Nitroprusside Dihydrate, puriss. p.a., ≥99%, ACS reagent, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.