missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Persulfate, Honeywell
Reagent Grade,≥98%
36.54 € - 296.38 €
Chemical Identifiers
CAS | 7775-27-1 |
---|---|
Molecular Formula | Na2O8S2 |
Molecular Weight (g/mol) | 238.092 |
MDL Number | MFCD00003501 |
InChI Key | CHQMHPLRPQMAMX-UHFFFAOYSA-L |
Synonym | sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate |
PubChem CID | 62655 |
IUPAC Name | disodium;sulfonatooxy sulfate |
SMILES | [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15632030
|
Honeywell Research Chemicals
216232-500G |
500 g | |||||||
15602030
|
Honeywell Research Chemicals
216232-1KG |
1 kg | |||||||
15612030
|
Honeywell Research Chemicals
216232-2.5KG |
2.5 kg | |||||||
15642030
|
Honeywell Research Chemicals
216232-5KG |
5 kg | |||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7775-27-1 | |
238.092 | |
CHQMHPLRPQMAMX-UHFFFAOYSA-L | |
62655 | |
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+] |
Na2O8S2 | |
MFCD00003501 | |
sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate | |
disodium;sulfonatooxy sulfate |
Specifications
7775-27-1 | |
Na2S2O8 | |
1505 | |
CHQMHPLRPQMAMX-UHFFFAOYSA-L | |
disodium;sulfonatooxy sulfate | |
62655 | |
≥98% | |
Sodium Persulfate |
Na2O8S2 | |
MFCD00003501 | |
sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate | |
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+] | |
238.092 | |
238.1g/mol | |
Reagent |
Safety and Handling
P220-P261-P280-P305 + P351 + P338-P342 + P311
H272-H302-H315-H317-H319-H334-H335
missing translation for 'einecsNumber' : 231-892-1
missing translation for 'rtecsNumber' : SE0525000