missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium phosphate, 0.5M buffer soln., pH 7.5

Brand:  Thermo Scientific Chemicals J61561.AK

96.20 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15454149

  • 96.68 € / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Sodium phosphate buffer is used routinely in the life sciences. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phosphate buffer is used routinely in the life sciences. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

Solubility
Soluble in water (121 mg/ml at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong acids.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sodium phosphate buffer soln.
7.5
250 mL
Soluble in water (121mg/ml at 20°C).
0.5 M
Liquid
MFCD00146206
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN25L506 Certificate TypeCertificate of Analysis Date09/04/2025 Catalog NumberJ61561.AK,J61561.AP
Lot NumberN25L505 Certificate TypeCertificate of Analysis Date14/03/2025 Catalog NumberJ61561.AK,J61561.AP
Lot NumberX28K532 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog NumberJ61561.AK,J61561.AP
Lot NumberW23K504 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog NumberJ61561.AK,J61561.AP
Lot NumberU24K504 Certificate TypeCertificate of Analysis Date30/09/2024 Catalog NumberJ61561.AK,J61561.AP
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Sodium phosphate, 0.5M buffer soln., pH 7.5 > 250mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only