missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium phosphate, 0.5M buffer soln., pH 7.5

96.68 € - 170.85 €

Specifications

Chemical Name or Material Sodium phosphate buffer soln.
Concentration 0.5 M
pH 7.5
Physical Form Liquid
MDL Number MFCD00146206
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15454149
View Documents
Thermo Scientific Chemicals
J61561.AK
250 mL
96.68 €
250mL
Please to purchase this item. Need a web account? Register with us today!
15464149
View Documents
Thermo Scientific Chemicals
J61561.AP
500 mL
170.85 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sodium phosphate buffer is used routinely in the life sciences. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phosphate buffer is used routinely in the life sciences. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

Solubility
Soluble in water (121 mg/ml at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong acids.
Specifications

Specifications

Sodium phosphate buffer soln.
7.5
MFCD00146206
0.5 M
Liquid
Soluble in water (121mg/ml at 20°C).
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only