missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate dibasic, Puriss., meets analytical spec. of BP, Ph. Eur., USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance), Honeywell Fluka™
Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
53.92 € - 956.92 €
Chemical Identifiers
CAS | 7558-79-4 |
---|---|
Molecular Formula | HNa2O4P |
Molecular Weight (g/mol) | 141.96 |
MDL Number | MFCD00003496 |
InChI Key | BNIILDVGGAEEIG-UHFFFAOYSA-L |
Synonym | disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt |
PubChem CID | 24203 |
ChEBI | CHEBI:34683 |
IUPAC Name | disodium;hydrogen phosphate |
SMILES | [Na+].[Na+].OP([O-])([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15670750
|
Honeywell-Fluka
04276-1KG |
1 kg |
53.92 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15600760
|
Honeywell-Fluka
04276-5KG |
5 kg |
285.26 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15610760
|
Honeywell-Fluka
04276-6X1KG |
6 x 1 kg |
252.32 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15690750
|
Honeywell-Fluka
04276-4X5KG |
4 x 5 kg |
956.92 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15680750
|
Honeywell-Fluka
04276-25KG |
25 kg |
610.25 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7558-79-4 | |
141.96 | |
BNIILDVGGAEEIG-UHFFFAOYSA-L | |
24203 | |
disodium;hydrogen phosphate |
HNa2O4P | |
MFCD00003496 | |
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
CHEBI:34683 | |
[Na+].[Na+].OP([O-])([O-])=O |
Specifications
7558-79-4 | |
HNa2O4P | |
MFCD00003496 | |
Very Soluble in hot water, 8 parts soluble in water , insoluble in alcohol | |
[Na+].[Na+].OP([O-])([O-])=O | |
141.96 | |
CHEBI:34683 | |
98 to 100.5% (calc. to the dried substance) | |
Sodium Phosphate Dibasic, Meets analytical specification of Ph.Eur.BP, USP, FCC |
Anhydrous | |
Na2HPO4 | |
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
BNIILDVGGAEEIG-UHFFFAOYSA-L | |
disodium;hydrogen phosphate | |
24203 | |
141.96g/mol | |
puriss. |
Safety and Handling
missing translation for 'einecsNumber' : 231-448-7
missing translation for 'rtecsNumber' : WC4500000