missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, monobasic monohydrate, 99+%, for analysis, Thermo Scientific Chemicals

39.50 € - 245.00 €

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
PubChem CID 516949
SMILES O.[Na+].OP(O)(O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10513544
View Documents
Thermo Scientific Acros
207802500
250 g Plastic bottle
39.50 €
250g
Please to purchase this item. Need a web account? Register with us today!
10226410
View Documents
Thermo Scientific Acros
207805000
500 g Plastic bottle
58.10 €
500g
Please to purchase this item. Need a web account? Register with us today!
10523864
View Documents
Thermo Scientific Acros
207800025
2.5 kg Plastic bottle
245.00 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

10049-21-5
139.00
BBMHARZCALWXSL-UHFFFAOYSA-N
516949
H5NaO5P
MFCD00149208
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
O.[Na+].OP(O)(O)=O

Specifications

100.0°C
White
Crystalline Powder and/or Crystals
H5NaO5P
MFCD00149208
15, 8790
Solubility in water: 1103g/L (20°C). Other solubilities: practically insoluble in alcohol
O.[Na+].OP(O)(O)=O
139.00
137.99
Analytical
10049-21-5
4.1 to 4.5 (5% soln. at 20°C)
99+%
NaH2PO4·H2O
01,395
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707
BBMHARZCALWXSL-UHFFFAOYSA-N
sodium;dihydrogen phosphate;hydrate
516949
≥99%
Sodium phosphate, monobasic monohydrate, p.a.
Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

Product Identifier
  • Sodium phosphate, monobasic monohydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only