missing translation for 'onlineSavingsMsg'
Learn More
Sodium Solution for ICP, 10g/L, HNO3 Matrix, Fisher Chemical™ Product Code.: 12967203

Sodium Solution for ICP, 10g/L, HNO3 Matrix, Fisher Chemical™
BIOPHARMA

Product Code. 12967203
100 mL
Click to view available options
Quantity:
100 mL
250 mL
500 mL
This item is not returnable. View return policy

Product Code. 12967203

Brand: Fisher Chemical S 335 100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Quantity 100 mL
Solution Contains HNO3
Concentration 10 g/L

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1097789 Certificate TypeCertificate of Analysis Date30/03/2025 Catalog NumberS 335 100
Lot Number1064213 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberS 335 100
Lot Number1053431 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberS 335 100
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Solution for ICP, 10g/L, HNO3 Matrix, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.