Learn More
Sodium sulfate, 99+%, ACS reagent, granular, anhydrous, Thermo Scientific Chemicals
45.10 € - 285.42 €
Chemical Identifiers
CAS | 7757-82-6 |
---|---|
Molecular Formula | Na2O4S |
Molecular Weight (g/mol) | 142.04 |
MDL Number | MFCD00003504 |
InChI Key | PMZURENOXWZQFD-UHFFFAOYSA-L |
Synonym | sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 |
PubChem CID | 24436 |
ChEBI | CHEBI:32149 |
SMILES | [Na+].[Na+].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10041501
|
Thermo Scientific Acros
354250010 |
1 kg | Plastic bottle |
45.10 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10504064
|
Thermo Scientific Acros
354250025 |
2.5 kg | Plastic bottle |
97.79 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13468528
|
Thermo Scientific Acros
354250100 |
10 kg | Plastic drum |
285.42 €
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7757-82-6 | |
142.04 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
24436 | |
[Na+].[Na+].[O-]S([O-])(=O)=O |
Na2O4S | |
MFCD00003504 | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
CHEBI:32149 |
Specifications
>1700.0°C | |
7757-82-6 | |
5.2 to 7.0 (5% soln. at 25°C) | |
99+% | |
Na2SO4 | |
01,1103 | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
[Na+].[Na+].[O-]S([O-])(=O)=O | |
142.04 | |
CHEBI:32149 | |
≥99% | |
Sodium : passes test (USP) Sulfate : passes test (USP) | |
Decomposition Information: >890°C | |
Sodium sulfate, Anhydrous |
884.0°C | |
Colorless to Orange | |
Granules | |
Na2O4S | |
MFCD00003504 | |
15, 8809 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
disodium sulfate | |
24436 | |
142.04 | |
ACS Reagent | |
0.5 % max. | |
2.68g/mL |
Safety and Handling
Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health: 1
Flammability: 1
Instability: 0
SIGNAL WORD: CAUTION!
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.