missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, 99+%, for analysis, crystalline, anhydrous, Thermo Scientific Chemicals

35.60 € - 205.00 €

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium;sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10206660
View Documents Promotion Details
Thermo Scientific Acros
219260010
1 kg Plastic bottle
35.60 €
1kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10022850
View Documents Promotion Details
Thermo Scientific Acros
219260025
2.5 kg Plastic bottle
73.00 €
2.50kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10404405
View Documents Promotion Details
Thermo Scientific Acros
219260100
10 kg Plastic drum
205.00 €
10kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7757-82-6
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
24436
disodium;sulfate
Na2O4S
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
CHEBI:32149
[Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

>1700.0°C
7757-82-6
5 to 8 (5% soln. at 20°C)
99+%
Na2SO4
01,1103
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium;sulfate
24436
142.04
Analytical
2.68g/mL
884.0°C
White
Crystalline Powder and/or Crystals
Na2O4S
MFCD00003504
15, 8809
Solubility in water: 185g/L. Other solubilities: soluble in glycerol and hydrogen iodide, insoluble in alcohol
[Na+].[Na+].[O-]S([O-])(=O)=O
142.04
CHEBI:32149
≥99%
0.5% max.
Sodium sulfate, Anhydrous
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-820-9

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only